Product Details

SNP ID
rs71390884
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:87329606 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAACAGTGACCACCACAGAGGG[C/T]AGGGAGTGACAAAGCTTGTAGGCTA
Phenotype
MIM: 609102
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXO31 PubMed Links

Gene Details

Gene
FBXO31
Gene Name
F-box protein 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282683.1 3351 UTR 3 NP_001269612.1
NM_024735.4 3351 UTR 3 NP_079011.3

View Full Product Details