Product Details

SNP ID
rs72827605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5478802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGGGTGGAAAAAACAATTATGG[C/T]ATACTAAGTTTACAAGGTTACAAAA
Phenotype
MIM: 610304 MIM: 614405 MIM: 609178
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DERL2 PubMed Links

Gene Details

Gene
DERL2
Gene Name
derlin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304777.1 Intron NP_001291706.1
NM_001304779.1 Intron NP_001291708.1
NM_016041.4 Intron NP_057125.2
Gene
DHX33
Gene Name
DEAH-box helicase 33
There are no transcripts associated with this gene.

Gene
LOC105371506
Gene Name
uncharacterized LOC105371506
There are no transcripts associated with this gene.

Gene
MIS12
Gene Name
MIS12, kinetochore complex component
There are no transcripts associated with this gene.

View Full Product Details