Product Details

SNP ID
rs72842820
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7425815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTCTTGTCAACATTGGCATCAAC[A/G]TGGCAACTATGGTCAGTGATGATTG
Phenotype
MIM: 606479 MIM: 615116 MIM: 613936
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C17orf74 PubMed Links

Gene Details

Gene
C17orf74
Gene Name
chromosome 17 open reading frame 74
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175734.4 200 Missense Mutation ATG,GTG M43V NP_783861.3
XM_006721471.3 200 UTR 5 XP_006721534.1
XM_011523722.2 200 UTR 5 XP_011522024.1
Gene
LOC107983988
Gene Name
proteoglycan 4-like
There are no transcripts associated with this gene.

Gene
NLGN2
Gene Name
neuroligin 2
There are no transcripts associated with this gene.

Gene
SPEM1
Gene Name
spermatid maturation 1
There are no transcripts associated with this gene.

Gene
TMEM102
Gene Name
transmembrane protein 102
There are no transcripts associated with this gene.

View Full Product Details