Product Details

SNP ID
rs73983173
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41140463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCACAGCCCTGGTCAGAGCAGA[C/G]GGAACCACAACAGGAGCTGACCATG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP4-5 PubMed Links

Gene Details

Gene
KRTAP4-5
Gene Name
keratin associated protein 4-5
There are no transcripts associated with this gene.

Gene
KRTAP4-6
Gene Name
keratin associated protein 4-6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030976.1 25 Missense Mutation CTC,GTC L9V NP_112238.1

View Full Product Details