Product Details

SNP ID
rs72837329
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:56795078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGATCCCCTTACACAGGATGAA[C/T]CCTGTTCCTGTCACAGTGGGGTTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C17orf67 PubMed Links

Gene Details

Gene
C17orf67
Gene Name
chromosome 17 open reading frame 67
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085430.2 1538 Missense Mutation ATT,GTT I87V NP_001078899.2
XM_011524735.2 1538 Intron XP_011523037.1

View Full Product Details