Product Details

SNP ID
rs72923678
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:50269647 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATATTTAACTCTGTGAGGAAGG[A/G]CACCAGCTTCTTCTGCAGGACACCC
Phenotype
MIM: 614759 MIM: 156535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP53 PubMed Links

Gene Details

Gene
CFAP53
Gene Name
cilia and flagella associated protein 53
There are no transcripts associated with this gene.

Gene
MBD1
Gene Name
methyl-CpG binding domain protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204136.1 2180 Silent Mutation TGC,TGT C650C NP_001191065.1
NM_001204137.1 2180 Intron NP_001191066.1
NM_001204138.1 2180 Intron NP_001191067.1
NM_001204139.1 2180 Intron NP_001191068.1
NM_001204140.1 2180 Intron NP_001191069.1
NM_001204141.1 2180 Intron NP_001191070.1
NM_001204142.1 2180 UTR 3 NP_001191071.1
NM_001204143.1 2180 UTR 3 NP_001191072.1
NM_001204151.2 2180 UTR 3 NP_001191080.1
NM_001323942.1 2180 Silent Mutation TGC,TGT C675C NP_001310871.1
NM_001323947.1 2180 Silent Mutation TGC,TGT C660C NP_001310876.1
NM_001323949.1 2180 UTR 3 NP_001310878.1
NM_001323950.1 2180 UTR 3 NP_001310879.1
NM_001323951.1 2180 Intron NP_001310880.1
NM_001323952.1 2180 UTR 3 NP_001310881.1
NM_001323953.1 2180 Intron NP_001310882.1
NM_001323954.1 2180 Silent Mutation TGC,TGT C571C NP_001310883.1
NM_002384.2 2180 UTR 3 NP_002375.1
NM_015844.2 2180 UTR 3 NP_056669.2
NM_015845.3 2180 Silent Mutation TGC,TGT C581C NP_056670.2
NM_015846.3 2180 UTR 3 NP_056671.2
NM_015847.3 2180 UTR 3 NP_056723.2
XM_005258271.2 2180 UTR 3 XP_005258328.1
XM_006722456.2 2180 UTR 3 XP_006722519.1
XM_011525991.1 2180 Silent Mutation TGC,TGT C629C XP_011524293.1
XM_011525993.2 2180 UTR 3 XP_011524295.1
XM_011525994.2 2180 UTR 3 XP_011524296.1
XM_011525998.2 2180 Intron XP_011524300.1
XM_011525999.1 2180 Silent Mutation TGC,TGT C604C XP_011524301.1
XM_011526001.1 2180 Silent Mutation TGC,TGT C594C XP_011524303.1
XM_011526002.1 2180 UTR 3 XP_011524304.1
XM_011526003.2 2180 UTR 3 XP_011524305.1
XM_011526006.1 2180 UTR 3 XP_011524308.1
XM_011526007.1 2180 UTR 3 XP_011524309.1
XM_017025751.1 2180 UTR 3 XP_016881240.1
XM_017025752.1 2180 Silent Mutation TGC,TGT C635C XP_016881241.1
XM_017025753.1 2180 UTR 3 XP_016881242.1
XM_017025754.1 2180 Intron XP_016881243.1
XM_017025755.1 2180 Intron XP_016881244.1
XM_017025756.1 2180 UTR 3 XP_016881245.1
XM_017025757.1 2180 UTR 3 XP_016881246.1
XM_017025758.1 2180 UTR 3 XP_016881247.1
XM_017025759.1 2180 UTR 3 XP_016881248.1
XM_017025760.1 2180 UTR 3 XP_016881249.1
XM_017025761.1 2180 Intron XP_016881250.1
XM_017025762.1 2180 UTR 3 XP_016881251.1
XM_017025763.1 2180 UTR 3 XP_016881252.1
XM_017025764.1 2180 UTR 3 XP_016881253.1
XM_017025765.1 2180 UTR 3 XP_016881254.1
XM_017025766.1 2180 UTR 3 XP_016881255.1
XM_017025767.1 2180 UTR 3 XP_016881256.1
XM_017025768.1 2180 Intron XP_016881257.1
XM_017025769.1 2180 UTR 3 XP_016881258.1
XM_017025770.1 2180 UTR 3 XP_016881259.1
XM_017025771.1 2180 UTR 3 XP_016881260.1
XM_017025772.1 2180 Intron XP_016881261.1
XM_017025773.1 2180 UTR 3 XP_016881262.1
XM_017025774.1 2180 UTR 3 XP_016881263.1
XM_017025775.1 2180 Intron XP_016881264.1
XM_017025776.1 2180 UTR 3 XP_016881265.1
XM_017025777.1 2180 Intron XP_016881266.1

View Full Product Details