Product Details

SNP ID
rs73043012
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38336812 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGGCGGAATAAGGAGCAAGGTG[A/G]GGATAAAGTTAAAGGAAGAGACGTG
Phenotype
MIM: 613452 MIM: 603939
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CATSPERG PubMed Links
Additional Information
For this assay, SNP(s) [rs72358859] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CATSPERG
Gene Name
cation channel sperm associated auxiliary subunit gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021185.4 Intron NP_067008.3
XM_005259114.4 Intron XP_005259171.1
XM_005259115.4 Intron XP_005259172.1
XM_005259116.2 Intron XP_005259173.1
XM_006723308.3 Intron XP_006723371.1
XM_006723310.2 Intron XP_006723373.1
XM_006723311.2 Intron XP_006723374.1
XM_011527174.2 Intron XP_011525476.1
XM_011527175.2 Intron XP_011525477.1
XM_011527176.2 Intron XP_011525478.1
XM_011527177.2 Intron XP_011525479.1
XM_011527178.2 Intron XP_011525480.1
XM_011527179.1 Intron XP_011525481.1
XM_011527182.2 Intron XP_011525484.1
XM_017027053.1 Intron XP_016882542.1
XM_017027054.1 Intron XP_016882543.1
XM_017027055.1 Intron XP_016882544.1
Gene
KCNK6
Gene Name
potassium two pore domain channel subfamily K member 6
There are no transcripts associated with this gene.

View Full Product Details