Product Details

SNP ID
rs73933660
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41555657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACATGCCACAAGACCAAGCCACA[A/C]AATTTTTCCACACTGCACAATGCTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CEACAM21 PubMed Links

Gene Details

Gene
CEACAM21
Gene Name
carcinoembryonic antigen related cell adhesion molecule 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098506.3 761 Intron NP_001091976.3
NM_001288773.2 761 Intron NP_001275702.2
NM_001290113.1 761 Intron NP_001277042.1
NM_033543.5 761 Intron NP_291021.4
XM_005278396.4 761 Intron XP_005278453.1
XM_005278397.3 761 Intron XP_005278454.1
XM_011527450.1 761 Intron XP_011525752.1
XM_011527452.1 761 Intron XP_011525754.1
XM_017027430.1 761 Intron XP_016882919.1
XM_017027431.1 761 Intron XP_016882920.1
XM_017027432.1 761 Intron XP_016882921.1
XM_017027433.1 761 Intron XP_016882922.1
XM_017027434.1 761 Intron XP_016882923.1
XM_017027435.1 761 Intron XP_016882924.1
XM_017027436.1 761 UTR 5 XP_016882925.1
XM_017027437.1 761 Intron XP_016882926.1

View Full Product Details