Product Details

SNP ID
rs73932697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50977674 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCAGGGTACCTCTGCACACCAA[A/C]GGTCCCCCTGAGTCACCCTAGAGGG
Phenotype
MIM: 602652 MIM: 604438
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KLK6 PubMed Links

Gene Details

Gene
KLK6
Gene Name
kallikrein related peptidase 6
There are no transcripts associated with this gene.

Gene
KLK7
Gene Name
kallikrein related peptidase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207053.1 896 Silent Mutation NP_001193982.1
NM_001243126.1 896 Silent Mutation NP_001230055.1
NM_005046.3 896 Silent Mutation NP_005037.1
NM_139277.2 896 Silent Mutation NP_644806.1

View Full Product Details