Product Details
- SNP ID
-
rs73561444
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:57746818 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGACTTCCGGCGTCCTCTACTAGT[A/G]GCCATTTTGATTGGTGTTGGGTGTA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF776
PubMed Links
Gene Details
- Gene
- ZNF776
- Gene Name
- zinc finger protein 776
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173632.3 |
23 |
UTR 5 |
|
|
NP_775903.3 |
View Full Product Details