Product Details

SNP ID
rs72985486
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2238731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAAAGCTCCCACCTCTCACACAC[C/T]TATCAGTTGTATTTTGCATAGGTTT
Phenotype
MIM: 607375 MIM: 600796
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DOT1L PubMed Links

Gene Details

Gene
DOT1L
Gene Name
DOT1 like histone lysine methyltransferase
There are no transcripts associated with this gene.

Gene
MIR1227
Gene Name
microRNA 1227
There are no transcripts associated with this gene.

Gene
MIR6789
Gene Name
microRNA 6789
There are no transcripts associated with this gene.

Gene
PLEKHJ1
Gene Name
pleckstrin homology domain containing J1
There are no transcripts associated with this gene.

Gene
SF3A2
Gene Name
splicing factor 3a subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007165.4 Intron NP_009096.2

View Full Product Details