Product Details
- SNP ID
-
rs72987172
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:5819174 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCATTCAAATGCTGGCTCTATGA[C/T]CTTGGACAAGTCACTTCCCACCTCT
- Phenotype
-
MIM: 602018
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NRTN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143077610] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NRTN
- Gene Name
- neurturin
View Full Product Details