Product Details
- SNP ID
-
rs72997346
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:18652055 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGCAGAGAAGGCTCTGGGGTGGAG[A/G]CAGCAGAGGTGCTGAGTGCAGAAAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KLHL26
PubMed Links
Gene Details
- Gene
- KLHL26
- Gene Name
- kelch like family member 26
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018316.1 |
|
Intron |
|
|
NP_060786.1 |
View Full Product Details