Product Details

SNP ID
rs73913917
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:56372917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTATCCCTACTATTGACAGAGGCA[A/G]ACTGAAGAGAATCTCTCACCACTGC
Phenotype
MIM: 603072
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AURKA PubMed Links

Gene Details

Gene
AURKA
Gene Name
aurora kinase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323303.1 Intron NP_001310232.1
NM_001323304.1 Intron NP_001310233.1
NM_001323305.1 Intron NP_001310234.1
NM_003600.3 Intron NP_003591.2
NM_198433.2 Intron NP_940835.1
NM_198434.2 Intron NP_940836.1
NM_198435.2 Intron NP_940837.1
NM_198436.2 Intron NP_940838.1
NM_198437.2 Intron NP_940839.1
XM_017028034.1 Intron XP_016883523.1
XM_017028035.1 Intron XP_016883524.1
Gene
FAM210B
Gene Name
family with sequence similarity 210 member B
There are no transcripts associated with this gene.

View Full Product Details