Product Details
- SNP ID
-
rs73901167
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:29005922 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCAGTCATCACTTTGGTGTTTAT[A/G]GCAGTAAGGGGGGGAGGAGGTGGCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RWDD2B
PubMed Links
Gene Details
- Gene
- RWDD2B
- Gene Name
- RWD domain containing 2B
View Full Product Details