Product Details

SNP ID
rs73906147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:39176409 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACAAAACAGCCCAGCCCCTGCCC[A/G]TGGGAACTCATCTCTGAGTTAAGTG
Phenotype
MIM: 605296
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRWD1 PubMed Links

Gene Details

Gene
BRWD1
Gene Name
bromodomain and WD repeat domain containing 1
There are no transcripts associated with this gene.

Gene
PSMG1
Gene Name
proteasome assembly chaperone 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261824.1 Intron NP_001248753.1
NM_001320795.1 Intron NP_001307724.1
NM_003720.3 Intron NP_003711.1
NM_203433.2 Intron NP_982257.1
XM_017028488.1 Intron XP_016883977.1

View Full Product Details