Product Details

SNP ID
rs73152516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:24559516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCCTTTGTCATCCCATCCGTAA[C/T]GTGAAGGTGTCTAACCAGATAACCT
Phenotype
MIM: 601062
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GUCD1 PubMed Links

Gene Details

Gene
GUCD1
Gene Name
guanylyl cyclase domain containing 1
There are no transcripts associated with this gene.

Gene
SNRPD3
Gene Name
small nuclear ribonucleoprotein D3 polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278656.1 Intron NP_001265585.1
NM_004175.4 Intron NP_004166.1

View Full Product Details