Product Details
- SNP ID
-
rs72619562
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:44248545 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTGAAAGTGGATGGATTTCTAGAA[C/T]GCTCCTGCCTGGGTTTCCATGGCCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA1644
PubMed Links
Gene Details
- Gene
- KIAA1644
- Gene Name
- KIAA1644
View Full Product Details