Product Details

SNP ID
rs73629052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:88749413 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGAGAAGCAGCCAGTCATACTAT[C/T]CTCAAATTTTGTGAGTTCAATTTAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPXCR1 PubMed Links

Gene Details

Gene
CPXCR1
Gene Name
CPX chromosome region, candidate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184771.1 241 UTR 5 NP_001171700.1
NM_033048.5 241 UTR 5 NP_149037.4

View Full Product Details