Product Details

SNP ID
rs73628812
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:27462580 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCGCATCGTTGGTCAAGAAGTC[C/T]CTATGCCTTTTTGGCTGATCCAAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP4R3CP PubMed Links

Gene Details

Gene
PPP4R3CP
Gene Name
protein phosphatase 4 regulatory subunit 3C, pseudogene
There are no transcripts associated with this gene.

View Full Product Details