Product Details

SNP ID
rs72663170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:102882245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGTTCCCATTCACTCGCTTTTTG[A/G]TAAATTTAAGCTCTTATGACTTCTA
Phenotype
MIM: 611507 MIM: 611527
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CISD2 PubMed Links

Gene Details

Gene
CISD2
Gene Name
CDGSH iron sulfur domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008388.4 Intron NP_001008389.1
Gene
SLC9B1
Gene Name
solute carrier family 9 member B1
There are no transcripts associated with this gene.

View Full Product Details