Product Details

SNP ID
rs72731860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:146708035 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGCTGAGTCTAGCCTTCCAGCC[A/G]TCTTGGCCAAGTCTCCAGCCTGTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTC29 PubMed Links

Gene Details

Gene
TTC29
Gene Name
tetratricopeptide repeat domain 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300761.2 Intron NP_001287690.1
NM_001317806.1 Intron NP_001304735.1
NM_031956.3 Intron NP_114162.2
XM_005263270.1 Intron XP_005263327.1
XM_006714335.1 Intron XP_006714398.1
XM_006714336.1 Intron XP_006714399.1
XM_006714339.2 Intron XP_006714402.1
XM_011532310.2 Intron XP_011530612.1

View Full Product Details