Product Details

SNP ID
rs73089418
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:209697669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTATGTACCTCTTTTGCCTTTTTG[A/G]GCTGTTTGTGGCCAGGATGCTGGGC
Phenotype
MIM: 600713
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSD11B1 PubMed Links

Gene Details

Gene
HSD11B1
Gene Name
hydroxysteroid 11-beta dehydrogenase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206741.1 Intron NP_001193670.1
NM_005525.3 Intron NP_005516.1
NM_181755.2 Intron NP_861420.1
Gene
LOC101930114
Gene Name
uncharacterized LOC101930114
There are no transcripts associated with this gene.

View Full Product Details