Product Details

SNP ID
rs72868124
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:10947667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGGAGTCTCCATGGAGGCTCAG[C/T]GTGAGGGCCTCTCAGAAAGCCCCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf127 PubMed Links

Gene Details

Gene
C1orf127
Gene Name
chromosome 1 open reading frame 127
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170754.1 2386 Missense Mutation CAC,CGC H823R NP_001164225.1
XM_017000373.1 2386 Missense Mutation CAC,CGC H787R XP_016855862.1

View Full Product Details