Product Details

SNP ID
rs72542426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:43838541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCAAGGGCTCTGCCTTACCTGA[A/C]GCTGTAGGAGGCATGGAGGATGCCC
Phenotype
MIM: 605459 MIM: 605460
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ABCG5 PubMed Links

Gene Details

Gene
ABCG5
Gene Name
ATP binding cassette subfamily G member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022436.2 300 Missense Mutation ATC,CTC I47L NP_071881.1
XM_005264480.3 300 Missense Mutation ATC,CTC I47L XP_005264537.1
XM_006712073.3 300 Missense Mutation ATC,CTC I47L XP_006712136.1
XM_006712074.3 300 Missense Mutation ATC,CTC I47L XP_006712137.1
XM_011533024.2 300 Missense Mutation ATC,CTC I47L XP_011531326.1
XM_011533025.2 300 UTR 5 XP_011531327.1
XM_011533026.2 300 Missense Mutation ATC,CTC I47L XP_011531328.1
XM_011533027.2 300 UTR 5 XP_011531329.1
XM_011533028.2 300 Intron XP_011531330.1
Gene
ABCG8
Gene Name
ATP binding cassette subfamily G member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022437.2 300 Intron NP_071882.1
XM_005264483.3 300 Intron XP_005264540.1
XM_011533029.2 300 Intron XP_011531331.1
XM_011533030.2 300 Intron XP_011531332.1

View Full Product Details