Product Details

SNP ID
rs73952062
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74153619 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATACAAAAACTGTAGTACAAGGA[A/C]GTTCAACTGTTTAAAAATCTGCACA
Phenotype
MIM: 613183 MIM: 609281
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BOLA3 PubMed Links

Gene Details

Gene
BOLA3
Gene Name
bolA family member 3
There are no transcripts associated with this gene.

Gene
BOLA3-AS1
Gene Name
BOLA3 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
MOB1A
Gene Name
MOB kinase activator 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317110.1 3890 UTR 3 NP_001304039.1
NM_001317111.1 3890 UTR 3 NP_001304040.1
NM_001317112.1 3890 UTR 3 NP_001304041.1
NM_018221.4 3890 UTR 3 NP_060691.2
XM_011532948.2 3890 UTR 3 XP_011531250.1

View Full Product Details