Product Details
- SNP ID
-
rs71337785
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:85343283 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTGCCCGGATCCTAGAATCAAGAT[T/C]CTTAAGGTCTGAGTACAAGTTCCGC
- Phenotype
-
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RETSAT
PubMed Links
Gene Details
- Gene
- RETSAT
- Gene Name
- retinol saturase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017750.3 |
1983 |
Missense Mutation |
AAT,GAT |
N598D |
NP_060220.3 |
View Full Product Details