Product Details

SNP ID
rs72847738
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:87961263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAGATTAACTTATAAATTCCAA[C/G]AGGAAAGCGCCTGAAAGCCACTGAC
Phenotype
MIM: 612705
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RGPD2 PubMed Links

Gene Details

Gene
RGPD2
Gene Name
RANBP2-like and GRIP domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001078170.2 Intron NP_001071638.2
XM_017004845.1 Intron XP_016860334.1
XM_017004846.1 Intron XP_016860335.1
XM_017004847.1 Intron XP_016860336.1
XM_017004848.1 Intron XP_016860337.1
XM_017004849.1 Intron XP_016860338.1
XM_017004850.1 Intron XP_016860339.1

View Full Product Details