Product Details

SNP ID
rs73024306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:8573765 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTTCCTTTGTTCTGACGTTGGT[A/G]TAACATTGCTTCTTGAAAAATGAAA
Phenotype
MIM: 610485 MIM: 604859
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00312 PubMed Links

Gene Details

Gene
LINC00312
Gene Name
long intergenic non-protein coding RNA 312
There are no transcripts associated with this gene.

Gene
LMCD1
Gene Name
LIM and cysteine rich domains 1
There are no transcripts associated with this gene.

View Full Product Details