Product Details

SNP ID
rs73814308
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9917800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAAGGGAGCAGAGCTGTCCCAGGC[C/T]CATGCCCTCCATGCCCACCTCAGCC
Phenotype
MIM: 610925 MIM: 614995
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
IL17RC PubMed Links

Gene Details

Gene
IL17RC
Gene Name
interleukin 17 receptor C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001203263.1 Intron NP_001190192.1
NM_001203264.1 Intron NP_001190193.1
NM_001203265.1 Intron NP_001190194.1
NM_032732.5 Intron NP_116121.2
NM_153460.3 Intron NP_703190.1
NM_153461.3 Intron NP_703191.1
XM_017007326.1 Intron XP_016862815.1
XM_017007327.1 Intron XP_016862816.1
XM_017007328.1 Intron XP_016862817.1
XM_017007329.1 Intron XP_016862818.1
XM_017007330.1 Intron XP_016862819.1
XM_017007331.1 Intron XP_016862820.1
XM_017007332.1 Intron XP_016862821.1
XM_017007333.1 Intron XP_016862822.1
XM_017007334.1 Intron XP_016862823.1
XM_017007335.1 Intron XP_016862824.1
XM_017007336.1 Intron XP_016862825.1
XM_017007337.1 Intron XP_016862826.1
XM_017007338.1 Intron XP_016862827.1
XM_017007339.1 Intron XP_016862828.1
XM_017007340.1 Intron XP_016862829.1
XM_017007341.1 Intron XP_016862830.1
XM_017007342.1 Intron XP_016862831.1
XM_017007343.1 Intron XP_016862832.1
XM_017007344.1 Intron XP_016862833.1
XM_017007345.1 Intron XP_016862834.1
XM_017007346.1 Intron XP_016862835.1
XM_017007347.1 Intron XP_016862836.1
XM_017007348.1 Intron XP_016862837.1
XM_017007349.1 Intron XP_016862838.1
Gene
IL17RE
Gene Name
interleukin 17 receptor E
There are no transcripts associated with this gene.

View Full Product Details