Product Details
- SNP ID
-
rs73350301
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:178611679 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAACTACCAAGTCCCGTTGGACACA[C/T]AAAAAAAGTAAAAAGTATTAAAATA
- Phenotype
-
MIM: 607969
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CLK4
PubMed Links
Gene Details
- Gene
- CLK4
- Gene Name
- CDC like kinase 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020666.2 |
|
Intron |
|
|
NP_065717.1 |
View Full Product Details