Product Details
- SNP ID
-
rs73806855
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:179068500 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTAATAGGATTTAGTATGATTTACA[C/T]TGTGTTTTTGTTTTCAATTTAGACA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF354C
PubMed Links
Gene Details
- Gene
- ZNF354C
- Gene Name
- zinc finger protein 354C
View Full Product Details