Product Details

SNP ID
rs72816989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:180602616 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTGGCCACCCCAGGGGCTAGTT[A/G]GCTGTTTGGTCAGGCCCAGAAGAGG
Phenotype
MIM: 136352
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLT4 PubMed Links

Gene Details

Gene
FLT4
Gene Name
fms related tyrosine kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002020.4 5204 Intron NP_002011.2
NM_182925.4 5204 UTR 3 NP_891555.2
XM_011534478.2 5204 UTR 3 XP_011532780.1
XM_011534484.2 5204 UTR 3 XP_011532786.1
XM_017009263.1 5204 UTR 3 XP_016864752.1
XM_017009264.1 5204 Intron XP_016864753.1
XM_017009265.1 5204 Intron XP_016864754.1
XM_017009266.1 5204 Intron XP_016864755.1
XM_017009267.1 5204 Intron XP_016864756.1
XM_017009268.1 5204 UTR 3 XP_016864757.1

View Full Product Details