Product Details

SNP ID
rs73494140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:51759981 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACTTGTTCATACAATATAAAAG[A/C]AGCTATTTTTTTAACAAATGGTTTA
Phenotype
MIM: 616163 MIM: 610418
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DHRS12 PubMed Links

Gene Details

Gene
DHRS12
Gene Name
dehydrogenase/reductase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031719.2 1755 Intron NP_001026889.1
NM_001270424.1 1755 Intron NP_001257353.1
NM_024705.2 1755 Intron NP_078981.1
XM_005266527.3 1755 Intron XP_005266584.1
XM_005266528.4 1755 Intron XP_005266585.1
XM_011535235.2 1755 Intron XP_011533537.1
XM_011535236.2 1755 Intron XP_011533538.1
XM_011535237.1 1755 Intron XP_011533539.1
XM_011535238.1 1755 Intron XP_011533540.1
XM_017020749.1 1755 Intron XP_016876238.1
XM_017020750.1 1755 Intron XP_016876239.1
XM_017020751.1 1755 Intron XP_016876240.1
XM_017020752.1 1755 Intron XP_016876241.1
XM_017020753.1 1755 Intron XP_016876242.1
XM_017020754.1 1755 Intron XP_016876243.1
Gene
WDFY2
Gene Name
WD repeat and FYVE domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052950.3 1755 UTR 3 NP_443182.1
XM_011534914.1 1755 Intron XP_011533216.1
XM_011534915.2 1755 UTR 3 XP_011533217.1
XM_017020386.1 1755 UTR 3 XP_016875875.1

View Full Product Details