Product Details

SNP ID
rs73478206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:72538203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGAGGAAACACCAAGAAGCCCGA[A/G]AATGTGGAGTATTTCCAGCCCCCCA
Phenotype
MIM: 600433
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CBLN2 PubMed Links

Gene Details

Gene
CBLN2
Gene Name
cerebellin 2 precursor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182511.3 1718 Silent Mutation TTC,TTT F216F NP_872317.1
XM_006722394.3 1718 Silent Mutation TTC,TTT F216F XP_006722457.1
XM_011525824.2 1718 Silent Mutation TTC,TTT F216F XP_011524126.1
XM_017025559.1 1718 Silent Mutation TTC,TTT F216F XP_016881048.1
XM_017025560.1 1718 Silent Mutation TTC,TTT F216F XP_016881049.1

View Full Product Details