Product Details
- SNP ID
-
rs72965123
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:116580445 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTAGGCATTATGCTGGATATTGTC[A/G]TGTGGAGGTGCTAAGGGGCTGTTAC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RWDD1
PubMed Links
Gene Details
- Gene
- RWDD1
- Gene Name
- RWD domain containing 1
View Full Product Details