Product Details

SNP ID
rs74003538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:25054091 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTCCTCTCCTAGAGTGGATAGTC[G/T]GTGCTCATTGCTCAGTGGCATCGCT
Phenotype
MIM: 605436
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SNORD116-1 PubMed Links

Gene Details

Gene
SNORD116-1
Gene Name
small nucleolar RNA, C/D box 116-1
There are no transcripts associated with this gene.

Gene
SNORD116-2
Gene Name
small nucleolar RNA, C/D box 116-2
There are no transcripts associated with this gene.

Gene
SNORD116-3
Gene Name
small nucleolar RNA, C/D box 116-3
There are no transcripts associated with this gene.

Gene
SNORD116-4
Gene Name
small nucleolar RNA, C/D box 116-4
There are no transcripts associated with this gene.

Gene
SNORD116-5
Gene Name
small nucleolar RNA, C/D box 116-5
There are no transcripts associated with this gene.

Gene
SNORD116@
Gene Name
small nucleolar RNA, C/D box 116 cluster
There are no transcripts associated with this gene.

View Full Product Details