Product Details

SNP ID
rs74163510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122849398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGCCCACCACACCTTGTCTGGG[C/T]TGTGATTTTTTACAGCCACAGCTTC
Phenotype
MIM: 616644
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CUZD1 PubMed Links

Gene Details

Gene
CUZD1
Gene Name
CUB and zona pellucida like domains 1
There are no transcripts associated with this gene.

Gene
FAM24B
Gene Name
family with sequence similarity 24 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204364.1 438 Missense Mutation AAC,AGC N45S NP_001191293.1
NM_152644.2 438 Missense Mutation AAC,AGC N45S NP_689857.2
Gene
FAM24B-CUZD1
Gene Name
FAM24B-CUZD1 readthrough
There are no transcripts associated with this gene.

View Full Product Details