Product Details
- SNP ID
-
rs74101453
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
13
- Location
-
Chr.1:93183834 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAAAGTAATTATGATTTATTTAAA[A/C]TTTAGTGGAAACATTGCCTTATGTT
- Phenotype
-
MIM: 616876
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC18
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs57274666] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC18
- Gene Name
- coiled-coil domain containing 18
- Gene
- TMED5
- Gene Name
- transmembrane p24 trafficking protein 5
There are no transcripts associated with this gene.
View Full Product Details