Product Details

SNP ID
rs73020470
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:117901054 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCAGCAGGGCTGCCTGGAAGGCA[C/T]GGGACCATGTTAGCTTTCTCTCCAT
Phenotype
MIM: 610050
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMPRSS13 PubMed Links

Gene Details

Gene
TMPRSS13
Gene Name
transmembrane protease, serine 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077263.2 2930 UTR 3 NP_001070731.1
NM_001206789.1 2930 UTR 3 NP_001193718.1
NM_001206790.1 2930 Intron NP_001193719.1
NM_001244995.1 2930 Intron NP_001231924.1

View Full Product Details