Product Details

SNP ID
rs72928167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62126067 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGAACAAAGACTACTGTGTCCTT[A/G]AATCAAGCCTGAGGAAGTCCCATGA
Phenotype
MIM: 604411
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INCENP PubMed Links

Gene Details

Gene
INCENP
Gene Name
inner centromere protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040694.1 Intron NP_001035784.1
NM_020238.2 Intron NP_064623.2
XM_006718533.2 Intron XP_006718596.1
XM_011544995.2 Intron XP_011543297.1
XM_011544996.2 Intron XP_011543298.1
XM_011544997.2 Intron XP_011543299.1
XM_011544998.2 Intron XP_011543300.1

View Full Product Details