Product Details

SNP ID
rs72951702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72085667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCATGCAGAAGTTAGCTATTGCC[A/G]TGTAATAAACCATCCCAAAACACAG
Phenotype
MIM: 612414 MIM: 164009
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRTOMT PubMed Links

Gene Details

Gene
LRTOMT
Gene Name
leucine rich transmembrane and O-methyltransferase domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145307.4 Intron NP_001138779.1
NM_001145308.4 Intron NP_001138780.1
NM_001145309.3 Intron NP_001138781.1
NM_001145310.3 Intron NP_001138782.1
NM_001205138.3 Intron NP_001192067.1
NM_001271471.2 Intron NP_001258400.1
NM_001318803.1 Intron NP_001305732.1
NM_145309.5 Intron NP_660352.1
XM_006718473.3 Intron XP_006718536.1
XM_006718474.3 Intron XP_006718537.1
XM_011544847.2 Intron XP_011543149.1
XM_011544848.2 Intron XP_011543150.1
XM_017017356.1 Intron XP_016872845.1
XM_017017357.1 Intron XP_016872846.1
XM_017017358.1 Intron XP_016872847.1
XM_017017359.1 Intron XP_016872848.1
XM_017017360.1 Intron XP_016872849.1
Gene
NUMA1
Gene Name
nuclear mitotic apparatus protein 1
There are no transcripts associated with this gene.

View Full Product Details