Product Details

SNP ID
rs73360364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:55955117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTACCTACCTGGAGAGTCCGTTGA[C/G]TTCGGAAACCACCCAGCTAAGCAAA
Phenotype
MIM: 603004 MIM: 611980
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GBAS PubMed Links

Gene Details

Gene
GBAS
Gene Name
glioblastoma amplified sequence
There are no transcripts associated with this gene.

Gene
MRPS17
Gene Name
mitochondrial ribosomal protein S17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015969.2 Intron NP_057053.1

View Full Product Details