Product Details

SNP ID
rs72760514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:99568215 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGGGTGTTCAGTGGAAGGAAAA[C/T]AGCTTATTGACACCCAGAAATAATT
Phenotype
MIM: 600660
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MEF2A PubMed Links

Gene Details

Gene
MEF2A
Gene Name
myocyte enhancer factor 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130926.2 1361 Intron NP_001124398.1
NM_001130927.2 1361 Intron NP_001124399.1
NM_001130928.2 1361 Intron NP_001124400.1
NM_001171894.2 1361 Intron NP_001165365.1
NM_001319206.1 1361 Intron NP_001306135.1
NM_005587.3 1361 Intron NP_005578.2
XM_005254914.2 1361 Intron XP_005254971.1
XM_005254915.2 1361 Intron XP_005254972.1
XM_005254916.3 1361 Intron XP_005254973.1
XM_011521571.2 1361 Intron XP_011519873.1
XM_011521572.2 1361 Intron XP_011519874.1
XM_011521573.2 1361 Intron XP_011519875.1
XM_011521576.2 1361 Intron XP_011519878.1
XM_011521577.2 1361 Intron XP_011519879.1
XM_011521578.2 1361 Intron XP_011519880.1
XM_011521579.2 1361 Intron XP_011519881.1
XM_011521581.2 1361 Intron XP_011519883.1
XM_011521582.2 1361 Intron XP_011519884.1
XM_011521583.2 1361 Intron XP_011519885.1
XM_011521585.2 1361 Intron XP_011519887.1
XM_011521586.2 1361 Intron XP_011519888.1
XM_011521587.2 1361 Intron XP_011519889.1
XM_011521590.2 1361 Intron XP_011519892.1
XM_017022190.1 1361 Intron XP_016877679.1
XM_017022191.1 1361 Intron XP_016877680.1
XM_017022192.1 1361 Intron XP_016877681.1
XM_017022193.1 1361 Intron XP_016877682.1
XM_017022194.1 1361 Intron XP_016877683.1
XM_017022195.1 1361 Intron XP_016877684.1
XM_017022196.1 1361 Intron XP_016877685.1
XM_017022197.1 1361 UTR 5 XP_016877686.1
XM_017022198.1 1361 Intron XP_016877687.1
XM_017022199.1 1361 Intron XP_016877688.1
XM_017022200.1 1361 Intron XP_016877689.1
XM_017022201.1 1361 Intron XP_016877690.1
XM_017022202.1 1361 Intron XP_016877691.1
XM_017022203.1 1361 Intron XP_016877692.1
XM_017022204.1 1361 Intron XP_016877693.1

View Full Product Details