Product Details

SNP ID
rs74120006
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:1162738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAACGTGCTGTTAGGATATCAGGG[C/T]CTGGGAGCTGCAGCAATGTGGGCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00200 PubMed Links

Gene Details

Gene
LINC00200
Gene Name
long intergenic non-protein coding RNA 200
There are no transcripts associated with this gene.

View Full Product Details