Product Details

SNP ID
rs73921563
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7763528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAGACAGAGCCTCCCAGACCCC[G/T]GGGTCCTGGGGAGGTAGGTGTTGGG
Phenotype
MIM: 605872
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLEC4M PubMed Links

Gene Details

Gene
CLEC4M
Gene Name
C-type lectin domain family 4 member M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144904.1 Intron NP_001138376.1
NM_001144905.1 Intron NP_001138377.1
NM_001144906.1 Intron NP_001138378.1
NM_001144907.1 Intron NP_001138379.1
NM_001144908.1 Intron NP_001138380.1
NM_001144909.1 Intron NP_001138381.1
NM_001144910.1 Intron NP_001138382.1
NM_001144911.1 Intron NP_001138383.1
NM_014257.4 Intron NP_055072.3
XM_006722611.2 Intron XP_006722674.1
XM_006722612.3 Intron XP_006722675.1
XM_006722613.3 Intron XP_006722676.1
XM_006722614.3 Intron XP_006722677.1
XM_006722615.2 Intron XP_006722678.1

View Full Product Details