Product Details

SNP ID
rs72552295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:38352698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCCTTTACACAATTAAAAGAAGA[C/T]GCTGTTTAATCTGAGGATCCTGTTA
Phenotype
MIM: 300461
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OTC PubMed Links

Gene Details

Gene
OTC
Gene Name
ornithine carbamoyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000531.5 171 Missense Mutation ACG,ATG T1M NP_000522.3
XM_017029556.1 171 Missense Mutation ACG,ATG T1M XP_016885045.1

View Full Product Details