Product Details

SNP ID
rs76549678
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:160832643 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCTCCCACTCCTAAGTGATGTG[A/G]GAGGTCCCAAAAGAGACTCAGGGCC
Phenotype
MIM: 605554 MIM: 600684
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD244 PubMed Links

Gene Details

Gene
CD244
Gene Name
CD244 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166663.1 1262 Intron NP_001160135.1
NM_001166664.1 1262 Intron NP_001160136.1
NM_016382.3 1262 Intron NP_057466.1
XM_011509621.2 1262 UTR 3 XP_011507923.1
XM_011509622.2 1262 UTR 3 XP_011507924.1
XM_011509623.2 1262 Intron XP_011507925.1
Gene
LY9
Gene Name
lymphocyte antigen 9
There are no transcripts associated with this gene.

View Full Product Details