Product Details

SNP ID
rs78851184
Assay Type
Functionally Tested
NCBI dbSNP Submissions
32
Location
Chr.1:162854763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTTATTGTCAGGCTATGTTAAAC[A/G]GTTAGAGAGGAAGAAACTTAGAAGG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CCDC190 PubMed Links

Gene Details

Gene
CCDC190
Gene Name
coiled-coil domain containing 190
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178550.4 3704 UTR 3 NP_848645.3
XM_005245125.3 3704 UTR 3 XP_005245182.1
XM_006711289.3 3704 UTR 3 XP_006711352.1
XM_011509477.2 3704 UTR 3 XP_011507779.1

View Full Product Details