Product Details
- SNP ID
-
rs77936519
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:150364812 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTCGTTGGATCGAAAATTCCAGT[C/T]GGTAACCAACACCATGGAGTCCATT
- Phenotype
-
MIM: 614695
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RPRD2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs72696841] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RPRD2
- Gene Name
- regulation of nuclear pre-mRNA domain containing 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001297673.1 |
377 |
Missense Mutation |
TCG,TTG |
S33L |
NP_001284602.1 |
NM_001297674.1 |
377 |
Missense Mutation |
TCG,TTG |
S33L |
NP_001284603.1 |
NM_015203.4 |
377 |
Missense Mutation |
TCG,TTG |
S33L |
NP_056018.2 |
XM_005245033.2 |
377 |
Missense Mutation |
TCG,TTG |
S33L |
XP_005245090.1 |
XM_006711240.2 |
377 |
Missense Mutation |
TCG,TTG |
S33L |
XP_006711303.1 |
XM_011509344.2 |
377 |
Missense Mutation |
TCG,TTG |
S33L |
XP_011507646.1 |
XM_017000763.1 |
377 |
Missense Mutation |
TCG,TTG |
S33L |
XP_016856252.1 |
XM_017000764.1 |
377 |
Intron |
|
|
XP_016856253.1 |
XM_017000765.1 |
377 |
Missense Mutation |
TCG,TTG |
S33L |
XP_016856254.1 |
XM_017000766.1 |
377 |
Missense Mutation |
TCG,TTG |
S33L |
XP_016856255.1 |
XM_017000767.1 |
377 |
Missense Mutation |
TCG,TTG |
S33L |
XP_016856256.1 |
View Full Product Details